siRNA Details
Gene Symbol: Fiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106223.1 | NP_001099693.1 | 3 | 1429 | ||
XM_006228252.2 | XP_006228314.1 | 3 | 1537 | |||
XM_006228253.2 | XP_006228315.1 | 3 | 1384 | |||
XM_006228254.2 | XP_006228316.1 | 3 | 1969 | |||
GenBank mRNA | FQ224110.1 | - | 3 | 1368 | ||
FQ224124.1 | - | 3 | 1368 |
