siRNA Details
Gene Symbol: Ggn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013065.1 | NP_001013083.1 | 1 | 1216 | ||
XM_006228718.2 | XP_006228780.1 | 2 | 1811 | |||
XM_006228719.2 | XP_006228781.1 | 2 | 1645 | |||
XM_008759123.1 | XP_008757345.1 | 2 | 1971 | |||
GenBank mRNA | BC081922.1 | - | 1 | 1216 | 7112701 | |
BC100147.1 | - | 2 | 960 | 7111711 |
