siRNA Details
Gene Symbol: Ggn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013065.1 | NP_001013083.1 | 1 | 337 | ||
XM_008759123.1 | XP_008757345.1 | 2 | 1092 | |||
GenBank mRNA | BC081922.1 | - | 1 | 337 | 7112701 |
