siRNA Details
Gene Symbol: Sult2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039665.1 | NP_001034754.1 | 3, 4 | 601 | ||
XM_006229031.2 | XP_006229093.1 | 3, 4 | 502 | |||
XM_006229032.2 | XP_006229094.1 | 3, 4 | 467 | |||
XM_006229033.2 | XP_006229095.1 | 3, 4 | 833 | |||
XM_008759338.1 | XP_008757560.1 | 3, 4 | 430 | |||
GenBank mRNA | AY827147.1 | - | 3, 4 | 601 | ||
AY827148.1 | - | 3, 4 | 470 | |||
BC127438.1 | - | 1275 |
