siRNA Details
Gene Symbol: Perp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106265.2 | NP_001099735.1 | 2, 3 | 460 | ||
GenBank mRNA | BC166608.1 | - | 2, 3 | 460 | 5599021 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Perp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106265.2 | NP_001099735.1 | 2, 3 | 460 | ||
GenBank mRNA | BC166608.1 | - | 2, 3 | 460 | 5599021 |