siRNA Details
Gene Symbol: Zfand6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007630.1 | NP_001007631.1 | 3, 4 | 381 | ||
XM_006229499.2 | XP_006229561.1 | 5, 6 | 686 | |||
XM_006229500.2 | XP_006229562.1 | 3, 4 | 366 | |||
XM_006229502.2 | XP_006229564.1 | 4, 5 | 509 | |||
XM_006229504.1 | XP_006229566.1 | 4, 5 | 520 | |||
XM_006229505.2 | XP_006229567.1 | 3, 4 | 374 | |||
XM_006229506.2 | XP_006229568.1 | 3, 4 | 371 | |||
XM_006229507.2 | XP_006229569.1 | 3, 4 | 377 | |||
XM_006229508.2 | XP_006229570.1 | 3, 4 | 372 | |||
XM_008759611.1 | XP_008757833.1 | 3, 4 | 1911 | |||
GenBank mRNA | BC076394.1 | - | 3, 4 | 381 | 7107784 |
