siRNA Details
Gene Symbol: Tmem9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106289.1 | NP_001099759.1 | 3, 4 | 470 | ||
XM_006229972.2 | XP_006230034.1 | 3, 4 | 1761 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106289.1 | NP_001099759.1 | 3, 4 | 470 | ||
XM_006229972.2 | XP_006230034.1 | 3, 4 | 1761 |