siRNA Details
Gene Symbol: Esrra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008511.2 | NP_001008511.2 | 5 | 822 | ||
XM_006230755.1 | XP_006230817.1 | 5 | 935 | |||
GenBank mRNA | AY280663.1 | - | 4 | 716 | ||
BC104701.1 | - | 5 | 822 |
