siRNA Details
Gene Symbol: Tnni1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017184.1 | NP_058880.1 | 1, 3 | 60 | ||
XM_006249865.2 | XP_006249927.1 | 3, 5 | 189 | |||
XM_006249866.2 | XP_006249928.1 | 3, 5 | 120 | |||
GenBank mRNA | FQ214889.1 | - | 3, 5 | 167 | ||
FQ215520.1 | - | 3, 5 | 142 | |||
FQ215574.1 | - | 3, 5 | 140 | |||
FQ215749.1 | - | 3, 5 | 140 | |||
FQ216094.1 | - | 3, 5 | 140 | |||
FQ216822.1 | - | 3, 5 | 140 | |||
FQ217411.1 | - | 3, 5 | 140 | |||
J04993.1 | - | 1, 3 | 60 |
