siRNA Details
Gene Symbol: Ptpmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105726.1 | NP_001099196.1 | 2 | 826 | ||
GenBank mRNA | FQ219112.1 | - | 2 | 466 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ptpmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105726.1 | NP_001099196.1 | 2 | 826 | ||
GenBank mRNA | FQ219112.1 | - | 2 | 466 |