siRNA Details
Gene Symbol: Hoga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106355.1 | NP_001099825.1 | 7 | 1152 | ||
XM_006231386.2 | XP_006231448.1 | 6 | 1150 | |||
GenBank mRNA | FQ210513.1 | - | 7 | 966 | ||
FQ218423.1 | - | 7 | 964 |
