siRNA Details
Gene Symbol: Col11a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212528.1 | NP_997693.1 | 60 | 4657 | ||
XM_006255928.2 | XP_006255990.1 | 64 | 5862 | |||
XM_006255929.2 | XP_006255991.1 | 61 | 5541 | |||
XM_008772720.1 | XP_008770942.1 | 66 | 5986 |
