siRNA Details
Gene Symbol: Col11a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212528.1 | NP_997693.1 | 4 | 800 | ||
XM_006255928.2 | XP_006255990.1 | 4 | 1648 | |||
XM_006255929.2 | XP_006255991.1 | 4 | 1648 | |||
XM_008772720.1 | XP_008770942.1 | 4 | 1649 | |||
XM_008772721.1 | XP_008770943.1 | 4 | 1754 |
Gene Symbol: LOC100911886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772788.1 | XP_008771010.1 | 4 | 528 |
