siRNA Details
Gene Symbol: Sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193568.1 | NP_001180497.1 | 9 | 1009 | ||
NM_001193569.1 | NP_001180498.1 | 9 | 947 | |||
NM_019232.3 | NP_062105.2 | 9 | 921 | |||
XM_006227723.1 | XP_006227785.1 | 11 | 1607 | |||
XM_008758606.1 | XP_008756828.1 | 9 | 1117 | |||
GenBank mRNA | BC078843.1 | - | 9 | 875 | 7130800 | |
L01624.1 | - | 9 | 926 |
