siRNA Details
Gene Symbol: Sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193568.1 | NP_001180497.1 | 4 | 436 | ||
NM_001193569.1 | NP_001180498.1 | 4 | 374 | |||
NM_019232.3 | NP_062105.2 | 4 | 348 | |||
XM_006227723.1 | XP_006227785.1 | 6 | 1034 | |||
XM_008758606.1 | XP_008756828.1 | 4 | 544 | |||
GenBank mRNA | BC078843.1 | - | 4 | 302 | 7130800 | |
FM042519.1 | - | 4 | 436 | |||
FM068981.1 | - | 4 | 354 | |||
FM123924.1 | - | 4 | 341 | |||
L01624.1 | - | 4 | 353 |
