siRNA Details
Gene Symbol: RGD1359334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007638.1 | NP_001007639.1 | 1 | 711 | ||
GenBank mRNA | BC083828.1 | - | 1 | 711 | 7112333 |

More Information
Other Species Targeted

