siRNA Details
Gene Symbol: Mab21l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106456.1 | NP_001099926.1 | 7 | 1411 | ||
XM_006233045.2 | XP_006233107.1 | 7 | 1322 | |||
XM_006233046.2 | XP_006233108.1 | 7 | 1606 | |||
XM_008761374.1 | XP_008759596.1 | 7 | 1303 | |||
XM_008761375.1 | XP_008759597.1 | 7 | 1227 |
