siRNA Details
Gene Symbol: Ccdc109b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233280.2 | XP_006233342.1 | 1266 | |||
XM_006233281.2 | XP_006233343.2 | 1760 | ||||
XM_008761542.1 | XP_008759764.1 | 4 | 493 |
