siRNA Details
Gene Symbol: LOC296235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037350.1 | NP_001032427.1 | 3 | 417 | ||
GenBank mRNA | BC101872.1 | - | 3 | 417 | 5621810 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC296235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037350.1 | NP_001032427.1 | 3 | 417 | ||
GenBank mRNA | BC101872.1 | - | 3 | 417 | 5621810 |