siRNA Details
Gene Symbol: Gabrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017289.1 | NP_058985.1 | 4 | 445 | ||
GenBank mRNA | BC087714.1 | - | 4 | 410 | 7317154 | |
M35162.1 | - | 4 | 445 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gabrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017289.1 | NP_058985.1 | 4 | 445 | ||
GenBank mRNA | BC087714.1 | - | 4 | 410 | 7317154 | |
M35162.1 | - | 4 | 445 |