siRNA Details
Gene Symbol: Cpne9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024982.1 | NP_001020153.1 | 21 | 1636 | ||
XM_006237022.2 | XP_006237084.1 | 20 | 2513 | |||
XM_008763190.1 | XP_008761412.1 | 20 | 2613 | |||
GenBank mRNA | BC091347.1 | - | 21 | 1636 | 7316601 | |
FQ213359.1 | - | 12 | 889 |
