siRNA Details
Gene Symbol: Gpr63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106640.1 | NP_001100110.1 | 2 | 569 | ||
XM_006237949.2 | XP_006238011.1 | 1 | 306 | |||
XM_008763592.1 | XP_008761814.1 | 2 | 820 | |||
XM_008763593.1 | XP_008761815.1 | 2 | 322 |
