siRNA Details
Gene Symbol: Nans | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106655.2 | NP_001100125.2 | 2 | 284 | ||
GenBank mRNA | BC161979.1 | - | 2 | 278 | 9029956 | |
CB704610.1 | - | 2 | 295 | |||
CV116524.1 | - | 1 | 40 |
