siRNA Details
Gene Symbol: Nans | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106655.2 | NP_001100125.2 | 5 | 733 | ||
GenBank mRNA | BC161979.1 | - | 5 | 727 | 9029956 | |
BG669394.1 | - | 1 | 90 | |||
CV116524.1 | - | 4 | 489 |
