siRNA Details
Gene Symbol: Trmt10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013090.2 | NP_001013108.1 | 5 | 621 | ||
XM_006238042.1 | XP_006238104.1 | 6 | 671 | |||
XM_006238043.1 | XP_006238105.1 | 6 | 870 | |||
XM_006238044.2 | XP_006238106.1 | 6 | 631 | |||
GenBank mRNA | BC086603.1 | - | 5 | 625 | 7315452 |
