siRNA Details
Gene Symbol: Prpf38a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106672.1 | NP_001100142.1 | 10 | 1126 | ||
GenBank mRNA | FQ221356.1 | - | 8 | 532 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prpf38a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106672.1 | NP_001100142.1 | 10 | 1126 | ||
GenBank mRNA | FQ221356.1 | - | 8 | 532 |