siRNA Details
Gene Symbol: Creb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013092.2 | NP_001013110.1 | 2 | 410 | ||
XM_006238050.1 | XP_006238112.1 | 2 | 436 | |||
XM_008763672.1 | XP_008761894.1 | 3 | 240 | |||
GenBank mRNA | BC062241.1 | - | 2 | 413 | 6887761 | |
FQ219084.1 | - | 2 | 388 | |||
KJ817427.1 | - | 2 | 254 |
