siRNA Details
Gene Symbol: Tcea3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015008.1 | NP_001015008.1 | 6, 7 | 685 | ||
GenBank mRNA | BC091180.1 | - | 6, 7 | 685 | 7325833 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tcea3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015008.1 | NP_001015008.1 | 6, 7 | 685 | ||
GenBank mRNA | BC091180.1 | - | 6, 7 | 685 | 7325833 |