siRNA Details
Gene Symbol: Agmat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048185.1 | NP_001041650.1 | 2, 3 | 501 | ||
XM_008764233.1 | XP_008762455.1 | 2, 3 | 693 | |||
GenBank mRNA | BC091231.1 | - | 2, 3 | 485 | ||
BC105628.1 | - | 2, 3 | 501 | 7322772 |
