siRNA Details
Gene Symbol: Agr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106725.1 | NP_001100195.1 | 4 | 443 | ||
XM_006240041.2 | XP_006240103.1 | 4 | 445 |

More Information
Other Species Targeted

Gene Ontology Categories:
