siRNA Details
Gene Symbol: Rpl10l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750173.3 | XP_003750221.1 | 1 | 415 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rpl10l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750173.3 | XP_003750221.1 | 1 | 415 |