siRNA Details
Gene Symbol: Flrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106750.1 | NP_001100220.1 | 2 | 2517 | ||
XM_006240416.2 | XP_006240478.1 | 1 | 2152 | |||
XM_008764760.1 | XP_008762982.1 | 2 | 2212 |
