siRNA Details
Gene Symbol: Fkbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013105.2 | NP_001013123.2 | 5 | 433 | ||
XM_006257344.2 | XP_006257406.1 | 4 | 1338 | |||
XM_008765695.1 | XP_008763917.1 | 4 | 336 | |||
XM_008765696.1 | XP_008763918.1 | 4 | 348 | |||
XM_008765697.1 | XP_008763919.1 | 4 | 325 | |||
GenBank mRNA | BC084709.1 | - | 3 | 1022 | 7301886 |
