siRNA Details
Gene Symbol: Arhgap42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242515.2 | XP_006242577.1 | 19, 20 | 2623 | ||
XM_008765909.1 | XP_008764131.1 | 19, 20 | 3076 | |||
XM_576354.6 | XP_576354.3 | 19, 20 | 2166 |
