siRNA Details
Gene Symbol: Olr1226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000442.1 | NP_001000442.1 | 674 | |||
GenBank mRNA | X89703.1 | - | 1 | 291 |
Gene Symbol: LOC100911438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242828.2 | XP_006242890.1 | 1 | 890 |
Gene Symbol: LOC100911758 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749653.3 | XP_003749701.2 | 1 | 890 |
