siRNA Details
Gene Symbol: Tex264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007665.1 | NP_001007666.1 | 4 | 707 | ||
XM_006243726.2 | XP_006243788.1 | 4 | 673 | |||
XM_006243727.2 | XP_006243789.1 | 4 | 974 | |||
XM_008766501.1 | XP_008764723.1 | 4 | 663 | |||
GenBank mRNA | BC083732.1 | - | 4 | 707 | 7191345 |
