siRNA Details
Gene Symbol: Cmtm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198754.2 | NP_942049.1 | 3, 4 | 791 | ||
XM_006243964.2 | XP_006244026.1 | 3, 4 | 497 | |||
GenBank mRNA | AF538323.1 | - | 3, 4 | 594 | ||
BC088094.1 | - | 3, 4 | 791 | 7325894 |
