siRNA Details
Gene Symbol: Arpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106919.1 | NP_001100389.1 | 6 | 545 | ||
XM_006245165.1 | XP_006245227.1 | 5 | 683 | |||
XM_006245166.1 | XP_006245228.1 | 6 | 500 | |||
GenBank mRNA | FQ214133.1 | - | 6 | 505 | ||
FQ231947.1 | - | 3 | 298 |
