siRNA Details
Gene Symbol: Mff | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039015.2 | NP_001034104.1 | 4 | 576 | ||
NM_001271284.1 | NP_001258213.1 | 4 | 576 | |||
NM_001276401.1 | NP_001263330.1 | 5 | 746 | |||
XM_006245195.1 | XP_006245257.1 | 3 | 484 | |||
XM_006245197.1 | XP_006245259.1 | 3 | 484 | |||
XM_006245198.1 | XP_006245260.1 | 3 | 484 | |||
XM_006245200.1 | XP_006245262.1 | 3 | 484 | |||
XM_006245201.1 | XP_006245263.1 | 3 | 484 | |||
XM_006245204.1 | XP_006245266.1 | 3 | 484 | |||
XM_006245205.1 | XP_006245267.1 | 3 | 484 | |||
XM_006245206.1 | XP_006245268.1 | 5 | 681 | |||
XM_006245208.1 | XP_006245270.1 | 3 | 484 | |||
XM_006245209.1 | XP_006245271.1 | 3 | 484 | |||
XM_006245211.1 | XP_006245273.1 | 3 | 484 | |||
XM_008767230.1 | XP_008765452.1 | 3 | 484 | |||
XM_008767231.1 | XP_008765453.1 | 5 | 681 | |||
XM_008767232.1 | XP_008765454.1 | 4 | 576 | |||
XM_008767233.1 | XP_008765455.1 | 4 | 576 | |||
GenBank mRNA | BC098682.1 | - | 3 | 371 | 7385142 | |
FM107137.1 | - | 2 | 82 | |||
FQ223111.1 | - | 5 | 666 | |||
FQ225115.1 | - | 4 | 525 |
Gene Symbol: LOC102556337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006245318.1 | XP_006245380.1 | 3 | 484 | ||
XM_006245320.1 | XP_006245382.1 | 3 | 484 | |||
XM_006245321.1 | XP_006245383.1 | 3 | 484 | |||
XM_006245323.1 | XP_006245385.1 | 3 | 484 | |||
XM_006245324.1 | XP_006245386.1 | 3 | 484 | |||
XM_006245327.1 | XP_006245389.1 | 3 | 484 | |||
XM_006245328.1 | XP_006245390.1 | 3 | 484 | |||
XM_006245329.1 | XP_006245391.1 | 5 | 681 | |||
XM_006245332.1 | XP_006245394.1 | 3 | 484 | |||
XM_006245334.1 | XP_006245396.1 | 3 | 484 | |||
XM_006245336.1 | XP_006245398.1 | 5 | 681 | |||
XM_008767321.1 | XP_008765543.1 | 3 | 484 | |||
XM_008767322.1 | XP_008765544.1 | 5 | 681 | |||
XM_008767323.1 | XP_008765545.1 | 4 | 576 | |||
XM_008767324.1 | XP_008765546.1 | 4 | 576 | |||
XM_008767325.1 | XP_008765547.1 | 4 | 576 |

More Information
Other Species Targeted

Gene Ontology Categories:
