siRNA Details
Gene Symbol: Fer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106928.1 | NP_001100398.1 | 12 | 1545 | ||
XM_006245600.2 | XP_006245662.1 | 17 | 2277 | |||
XM_008767367.1 | XP_008765589.1 | 17 | 3053 | |||
XM_008767369.1 | XP_008765591.1 | 18 | 2415 | |||
XM_008767370.1 | XP_008765592.1 | 17 | 3313 | |||
XM_008767371.1 | XP_008765593.1 | 17 | 3050 | |||
XM_008767372.1 | XP_008765594.1 | 10 | 1190 | |||
GenBank mRNA | X13412.1 | - | 7 | 631 |
