siRNA Details
Gene Symbol: Pof1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106934.1 | NP_001100404.1 | 14 | 1644 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pof1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106934.1 | NP_001100404.1 | 14 | 1644 |