siRNA Details
Gene Symbol: Tceal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009675.1 | NP_001009675.1 | 2 | 498 | ||
XM_006257294.1 | XP_006257356.1 | 2 | 563 | |||
XM_008773413.1 | XP_008771635.1 | 2 | 651 | |||
XM_008773414.1 | XP_008771636.1 | 2 | 512 | |||
GenBank mRNA | BC087575.1 | - | 2 | 498 | 7302183 | |
FQ214955.1 | - | 2 | 431 |
