siRNA Details
Gene Symbol: Amdhd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024990.2 | NP_001020161.2 | 5 | 613 | ||
XM_008767565.1 | XP_008765787.1 | 5 | 704 | |||
GenBank mRNA | BC091278.1 | - | 5 | 603 | ||
CO388545.1 | - | 5 | 613 |
