siRNA Details
Gene Symbol: Map2k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100674.1 | NP_001094144.1 | 9, 10 | 930 | ||
GenBank mRNA | BC100095.1 | - | 1, 2 | 49 | ||
BC160839.1 | - | 9, 10 | 800 | 7458293 | ||
FQ231052.1 | - | 3, 4 | 260 |
Gene Symbol: LOC103689956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767884.1 | XP_008766106.1 | 9, 10 | 984 |
