siRNA Details
Gene Symbol: Tmem98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007672.1 | NP_001007673.1 | 3 | 314 | ||
GenBank mRNA | BC078932.1 | - | 3 | 314 | 7107480 | |
FQ214419.1 | - | 3 | 333 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007672.1 | NP_001007673.1 | 3 | 314 | ||
GenBank mRNA | BC078932.1 | - | 3 | 314 | 7107480 | |
FQ214419.1 | - | 3 | 333 |