siRNA Details
Gene Symbol: Tbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107034.1 | NP_001100504.1 | 9 | 1351 | ||
XM_006247036.2 | XP_006247098.1 | 9 | 1452 | |||
XM_006247037.2 | XP_006247099.1 | 7 | 3374 | |||
XM_008768036.1 | XP_008766258.1 | 9 | 1241 | |||
XM_008768037.1 | XP_008766259.1 | 6 | 2053 |
