siRNA Details
Gene Symbol: Zfp385c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247466.2 | XP_006247528.1 | 2 | 1663 | ||
XM_008768216.1 | XP_008766438.1 | 3 | 386 | |||
XM_008768217.1 | XP_008766439.1 | 2 | 1663 | |||
XM_008768218.1 | XP_008766440.1 | 2 | 203 | |||
GenBank mRNA | BC162044.1 | - | 1 | 114 |
