siRNA Details
Gene Symbol: Cep97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107096.1 | NP_001100566.1 | 9 | 1120 | ||
XM_006248250.1 | XP_006248312.1 | 7 | 795 | |||
XM_008768642.1 | XP_008766864.1 | 9 | 1127 |
