siRNA Details
Gene Symbol: Ttyh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107124.1 | NP_001100594.1 | 5 | 723 | ||
XM_008768969.1 | XP_008767191.1 | 5 | 912 |

More Information
Other Species Targeted

Gene Ontology Categories:
