siRNA Details
Gene Symbol: Rilpl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191665.1 | NP_001178594.1 | 5 | 1031 | ||
XM_006249294.1 | XP_006249356.1 | 5 | 1295 | |||
XM_006249295.1 | XP_006249357.1 | 5 | 1295 | |||
XM_006249296.2 | XP_006249358.1 | 5 | 1295 | |||
XM_006249297.2 | XP_006249359.1 | 5 | 1295 |
